α,α,α-Trifluori-2-tolualdehydi ≥98%

Toimittaja: Thermo Fisher Scientific
Varoitus

B24591.06 B24591.14 B24591.22
B24591.06EA 78.8 EUR
B24591.06 B24591.14 B24591.22
α,α,α-Trifluori-2-tolualdehydi ≥98%
α,α,α-Trifluori-2-tolualdehydi

Purity: 98% CAS No.: 447-61-0 Molecular Formula: C8H5F3O Formula Weight: 174.12 Flash Point: 61°(141°F) Refractive Index: 1.4660 Odour: Almond-like Melting Point: -40° Boiling Point: 70-71°/16mm Beilstein Reference: 2045512 MDL No.: MFCD00003337

Kaava: C₈H₅F₃O
Kiehumispiste: 70…71 °C (16 mmHg)
Sulamispiste: –40 °C
Tiheys: 1,322
Leimahduspiste: 61 °C (141 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003337
CAS-numero: 447-61-0
EINECS: 207-185-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR