Benzyltrimethylammonium tribromide ≥98%

Toimittaja: Thermo Fisher Scientific
Varoitus

B24470.06 B24470.14
B24470.06EA 82.6 EUR
B24470.06 B24470.14
Benzyltrimethylammonium tribromide ≥98%
Benzyltrimethylammonium tribromide

Purity: 98% CAS No.: 111865-47-5 Molecular Formula: C6H5CH2N(CH3)3Br3 Formula Weight: 389.97 Melting Point: 99-101° MDL No.: MFCD00134423

Kaava: C₁₀H₁₆Br₃N
Sulamispiste: 99…101 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00134423
CAS-numero: 111865-47-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR