Natriumhydroksidi ≥97%, pearls

Toimittaja: Thermo Fisher Scientific
Vaara

Synonyms: Caustic soda

B24414.0E B24414.36
B24414.0EEA 146 EUR
B24414.0E B24414.36
Natriumhydroksidi ≥97%, pearls
Natriumhydroksidi

Purity: 97% CAS No.: 1310-73-2 Molecular Formula: NaOH Formula Weight: 40.00 Odour: Odourless pH: 14 Melting Point: 318° Boiling Point: 1390° Merck Reference: 14,8627 UN No.: 1823 MDL No.: MFCD00003548

Kaava: NaOH
Kiehumispiste: 1390 °C
Sulamispiste: 318 °C
Tiheys: 2,13
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003548
CAS-numero: 1310-73-2
EINECS: 215-185-5
UN: 1823
ADR: 8,II
REACH: 01-2119457892-27
Merck Index: 14,8627

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR