4-Cyanobenzoyl chloride ≥98%

Toimittaja: Thermo Fisher Scientific
Vaara

B24333.06 B24333.14 B24333.22
B24333.06EA 126 EUR
B24333.06 B24333.14 B24333.22
4-Cyanobenzoyl chloride ≥98%
4-Cyanobenzoyl chloride

Purity: 98% CAS No.: 6068-72-0 Molecular Formula: C8H4ClNO Formula Weight: 165.58 Odour: Irritating Melting Point: 68-70° Beilstein Reference: 386729 UN No.: 3261 MDL No.: MFCD00001822

Kaava: C₈H₄ClNO
Kiehumispiste: 150…151 °C (25 mmHg)
Sulamispiste: 68…70 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00001822
CAS-numero: 6068-72-0
EINECS: 228-005-5
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR