1,3-Benzenedisulphonyl chloride ≥98%

Toimittaja: Thermo Fisher Scientific
Vaara

B24044.06 B24044.14 B24044.22
B24044.06EA 159 EUR
B24044.06 B24044.14 B24044.22
1,3-Benzenedisulphonyl chloride ≥98%
1,3-Benzenedisulphonyl chloride

Purity: 98% CAS No.: 585-47-7 Molecular Formula: C6H4Cl2O4S2 Formula Weight: 275.13 Melting Point: 58-60° Beilstein Reference: 1118508 UN No.: 3261 MDL No.: MFCD00041879

Kaava: C₆H₄Cl₂O₄S₂
Kiehumispiste: 145 °C (1 mmHg)
Sulamispiste: 58…60 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00041879
CAS-numero: 585-47-7
EINECS: 209-556-0
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR