3-Fluori-p-anisaldehydi ≥98%

Toimittaja: Thermo Fisher Scientific
Varoitus

B23827.03 B23827.06
B23827.03EA 52.5 EUR
B23827.03 B23827.06
3-Fluori-p-anisaldehydi ≥98%
3-Fluori-p-anisaldehydi

Purity: 98% CAS No.: 351-54-2 Molecular Formula: C8H7FO2 Formula Weight: 154.14 Flash Point: 110°(230°F) Melting Point: 34-35° Boiling Point: 129-132°/11mm Beilstein Reference: 1942621 MDL No.: MFCD00003349

Kaava: C₈H₇FO₂
Kiehumispiste: 129…132 °C (11 mmHg)
Sulamispiste: 34…35 °C
Leimahduspiste: >110 °C (230 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003349
CAS-numero: 351-54-2
EINECS: 206-514-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR