2-Ethylbutyryl chloride ≥99%

Toimittaja: Thermo Fisher Scientific
Vaara

B23774.14 B23774.22
B23774.14EA 196 EUR
B23774.14 B23774.22
2-Ethylbutyryl chloride ≥99%
2-Ethylbutyryl chloride

Purity: 99% CAS No.: 2736-40-5 Molecular Formula: (CH3CH2)2CHCOCl Formula Weight: 134.61 Flash Point: 33°(91°F) Refractive Index: 1.4250 Odour: Acrid Boiling Point: 137-139° UN No.: 3094 MDL No.: MFCD00018812

Kaava: C₆H₁₁ClO
Kiehumispiste: 137…139 °C
Tiheys: 0,980
Leimahduspiste: 33 °C (91 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00018812
CAS-numero: 2736-40-5
EINECS: 220-359-9
UN: 2920
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR