2-Methyl-4-(trifluoromethoxy)aniline ≥97%

Toimittaja: Thermo Fisher Scientific
Vaara

B23706.03 B23706.06
B23706.03EA 69.3 EUR
B23706.03 B23706.06
2-Methyl-4-(trifluoromethoxy)aniline ≥97%
2-Methyl-4-(trifluoromethoxy)aniline

Purity: 97+% CAS No.: 86256-59-9 Molecular Formula: C8H8F3NO Formula Weight: 191.15 UN No.: 2810 MDL No.: MFCD01631541

Kaava: C₈H₈F₃NO
Storage Temperature: Huoneenlämpö
MDL numero: MFCD01631541
CAS-numero: 86256-59-9
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR