1-Dodecylpyridinium chloride hydrate 98%

Toimittaja: Thermo Fisher Scientific
Varoitus

B23492.14 B23492.22
B23492.14EA 141 EUR
B23492.14 B23492.22
1-Dodecylpyridinium chloride hydrate 98%
1-Dodecylpyridinium chloride hydrate

Purity: 98% CAS No.: 207234-02-4 Molecular Formula: C17H30ClN.xH2O Formula Weight: 283.89(anhy) Melting Point: 88-90° MDL No.: MFCD00150002

Kaava: C₁₇H₃₀ClN·xH₂O
Sulamispiste: 88…90 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00150002
CAS-numero: 207234-02-4
EINECS: 203-232-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR