Benzyltrimethylammonium dichloroiodate 95%

Toimittaja: Thermo Fisher Scientific
Varoitus

B22892.06 B22892.14
B22892.06EA 120 EUR
B22892.06 B22892.14
Benzyltrimethylammonium dichloroiodate 95%
Benzyltrimethylammonium dichloroiodate

Purity: 95% CAS No.: 114971-52-7 Molecular Formula: C10H16NICl2 Formula Weight: 348.06 Melting Point: ca. 125° Beilstein Reference: 4611949

Kaava: C₁₀H₁₆Cl₂IN
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00075259
CAS-numero: 114971-52-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR