1,3-Diaminoguanidine hydrochloride 98%

Toimittaja: Thermo Fisher Scientific
Varoitus

B22615.14 B22615.22
B22615.14EA 240 EUR
B22615.14 B22615.22
1,3-Diaminoguanidine hydrochloride 98%
1,3-Diaminoguanidine hydrochloride

Purity: 98% CAS No.: 36062-19-8 Molecular Formula: CH7N5.HCl Formula Weight: 125.56 Melting Point: ca 181° dec. Beilstein Reference: 3556702 MDL No.: MFCD00012948

Kaava: CH₇N₅
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00012948
CAS-numero: 36062-19-8
EINECS: 253-900-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR