Magnesium silicate 60-120 mesh kromatografiaan

Toimittaja: Thermo Fisher Scientific

B22490.0B B22490.30
B22490.0BEA 636 EUR
B22490.0B B22490.30
Magnesium silicate 60-120 mesh kromatografiaan
Magnesium silicate

Magnesium silicate, activated, is used in polyether adsorption and decolorization effect. Also used in chromatography.
Notes: Incompatible with strong acids and hydrogen fluoride.

Kaava: MgO₃Si
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00078249
CAS-numero: 1343-88-0
EINECS: 215-681-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR