(±)-3-Metyyli-1-pentanoli ≥98%

Toimittaja: Thermo Fisher Scientific
Vaara

B22356.06 B22356.14 B22356.22
B22356.06EA 101 EUR
B22356.06 B22356.14 B22356.22
(±)-3-Metyyli-1-pentanoli ≥98%
(±)-3-Metyyli-1-pentanoli

Purity: 98% CAS No.: 589-35-5 Molecular Formula: C6H14O Formula Weight: 102.18 Flash Point: 58°(136°F) Refractive Index: 1.4180 Boiling Point: 151-152° Beilstein Reference: 1718979 UN No.: 1987 MDL No.: MFCD00002937

Kaava: C₆H₁₄O
Kiehumispiste: 151…152 °C
Sulamispiste: –70 °C
Tiheys: 0,823
Leimahduspiste: 58 °C (136 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00002937
CAS-numero: 589-35-5
EINECS: 209-644-9
UN: 1987
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR