L-Aspartic acid monosodium salt monohydrate 99%

Toimittaja: Thermo Fisher Scientific

B22321.22 B22321.36
B22321.22EA 105 EUR
B22321.22 B22321.36
L-Aspartic acid monosodium salt monohydrate 99%
L-Aspartic acid monosodium salt monohydrate

Purity: 99% CAS No.: 323194-76-9 Molecular Formula: C4H6NNaO4.H2O Formula Weight: 173.11 (155.10anhy) MDL No.: MFCD00152960

Kaava: C₄H₆NNaO₄·H₂O
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00152960
CAS-numero: 323194-76-9
EINECS: 223-264-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR