3-(Methylthio)propyl isothiocyanate ≥98%

Toimittaja: Thermo Fisher Scientific
Vaara

B22150.09
B22150.09EA 121 EUR
B22150.09
3-(Methylthio)propyl isothiocyanate ≥98%
3-(Methylthio)propyl isothiocyanate

Purity: 98% CAS No.: 505-79-3 Molecular Formula: CH3S(CH2)3NCS Formula Weight: 147.25 Flash Point: 110°(230°F) Refractive Index: 1.564 Odour: Pungent Boiling Point: 254° UN No.: 2922 MDL No.: MFCD00209521

Kaava: C₅H₉NS₂
Kiehumispiste: 254 °C
Tiheys: 1,113
Leimahduspiste: 110 °C (230 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00209521
CAS-numero: 505-79-3
EINECS: 208-020-3
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR