BAPTA tetraethyl ester ≥98%

Toimittaja: Thermo Fisher Scientific

B21971.03 B21971.06
B21971.03EA 204 EUR
B21971.03 B21971.06
BAPTA tetraethyl ester ≥98%
BAPTA tetraethyl ester

Purity: 98+% CAS No.: 73630-07-6 Molecular Formula: C30H40N2O10 Formula Weight: 588.66 Melting Point: 98-102° MDL No.: MFCD00532663

Kaava: C₃₀H₄₀N₂O₁₀
Sulamispiste: 98…102 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00532663
CAS-numero: 73630-07-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR