Tris(2-hydroksietyyli)-1,3,5-triatsiinitrioni ≥97%

Toimittaja: Thermo Fisher Scientific
Varoitus

B21697.0B B21697.18 B21697.30
B21697.0BEA 1060 EUR
B21697.0B B21697.18 B21697.30
Tris(2-hydroksietyyli)-1,3,5-triatsiinitrioni ≥97%
Tris(2-hydroksietyyli)-1,3,5-triatsiinitrioni

Purity: 97% CAS No.: 839-90-7 Molecular Formula: C9H15N3O6 Formula Weight: 261.24 Melting Point: 136-140° MDL No.: MFCD00003549

Kaava: C₉H₁₅N₃O₆
Kiehumispiste: 527 °C (1013 hPa)
Sulamispiste: 136…140 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003549
CAS-numero: 839-90-7
EINECS: 212-660-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR