D-(+)-Sorbose ≥98%

Toimittaja: Thermo Fisher Scientific

B21208.02 B21208.77
B21208.02EA 873 EUR
B21208.02 B21208.77
D-(+)-Sorbose ≥98%
D-(+)-Sorbose

Purity: 98% CAS No.: 3615-56-3 Molecular Formula: C6H12O6 Formula Weight: 180.16 Melting Point: 163-165° MDL No.: MFCD00151095

Kaava: C₆H₁₂O₆
Kiehumispiste: 380 °C (1013 hPa)
Sulamispiste: 163…165 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00151095
CAS-numero: 3615-56-3
EINECS: 222-796-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR