(S)-(-)-1-Fenyylietanoli ≥98%

Toimittaja: Thermo Fisher Scientific
Vaara

B21188.03 B21188.06 B21188.14
B21188.03EA 73.6 EUR
B21188.03 B21188.06 B21188.14
(S)-(-)-1-Fenyylietanoli ≥98%
(S)-(-)-1-Fenyylietanoli

Purity: 99% CAS No.: 1445-91-6 Molecular Formula: C8H10O Formula Weight: 122.17 Flash Point: 85°(185°F) Refractive Index: 1.5280 Odour: Mild Melting Point: 9-11° Boiling Point: 97-99°/20mm Beilstein Reference: 2039797 Optical Rotation: -45° (c=5 in methanol) UN No.: 2937 MDL No.: MFCD00064264

Kaava: C₈H₁₀O
Kiehumispiste: 97…99 °C (20 mmHg)
Sulamispiste: 9…11 °C
Tiheys: 1,012
Leimahduspiste: 85 °C (185 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00064264
CAS-numero: 1445-91-6
UN: 2937
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR