5,6-Methylenedioxy-2-phenylindole 97%

Toimittaja: Thermo Fisher Scientific

B21065.06 B21065.03
B21065.06EA 560 EUR
B21065.06 B21065.03
5,6-Methylenedioxy-2-phenylindole 97%
5,6-Methylenedioxy-2-phenylindole

Purity: 97% CAS No.: 64943-90-4 Molecular Formula: C15H11NO2 Formula Weight: 237.26 MDL No.: MFCD00798602

Kaava: C₁₅H₁₁NO₂
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00798602
CAS-numero: 64943-90-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR