2,2-Difluori-1,3-bentsodioksoli ≥97%

Toimittaja: Thermo Fisher Scientific
Varoitus

B20540.06 B20540.14
B20540.06EA 135 EUR
B20540.06 B20540.14
2,2-Difluori-1,3-bentsodioksoli ≥97%
2,2-Difluori-1,3-bentsodioksoli

Purity: 97% CAS No.: 1583-59-1 Molecular Formula: C7H4F2O2 Formula Weight: 158.11 Flash Point: 32°(90°F) Refractive Index: 1.4430 Boiling Point: 129-130° Beilstein Reference: 1307648 UN No.: 1993 MDL No.: MFCD00236217

Kaava: C₇H₄F₂O₂
Kiehumispiste: 129…130 °C
Tiheys: 1,308
Leimahduspiste: 32 °C (90 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00236217
CAS-numero: 1583-59-1
EINECS: 216-431-4
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR