Di-n-propyl succinate 98%

Toimittaja: Thermo Fisher Scientific

B20441.09 B20441.18
B20441.09EA 53.2 EUR
B20441.09 B20441.18
Di-n-propyl succinate 98%
Di-n-propyl succinate

Purity: 98% CAS No.: 925-15-5 Molecular Formula: C10H18O4 Formula Weight: 202.25 Odour: Pleasant Boiling Point: 250-252° MDL No.: MFCD00015213

Kaava: C₁₀H₁₈O₄
Kiehumispiste: 250…252 °C
Tiheys: 1,00
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00015213
CAS-numero: 925-15-5
EINECS: 213-114-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR