Non return valves, Azlon®
Toimittaja: SciLabware
NWH902P NWH906P
AZLONWH902PEA 63.5 EUR
AZLONWH902P AZLONWH906P
Non return valves, Azlon®
Venttiilit Vastaventtiilit
Designed for use in vacuum lines and fluid handling systems.
  • Useful for preventing back pressure in filter flasks
  • Printed arrow indicates the direction of flow
  • Rigid with stepped tapered arms to give firm grip on tubing