Agarose LE, molecular biology grade, Axygen®
Toimittaja: Corning
AGR-LE-1001 AGR-LE-5001
AXYGAGR-LE-1001EA 1600 EUR
AXYGAGR-LE-1001 AXYGAGR-LE-5001
Agarose LE, molecular biology grade, Axygen®
Elektroforeesigeelit Agaroosi ja akryyliamidi
Axygen® Agarose Low Electroendosmosis (LE) is a high quality molecular biology grade agarose suitable for analytical and preparative electrophoresis of nucleic acids.
Separation range: 1 to 2,5 Kbp.