Anti-CCNA2 Rabbit Polyclonal Antibody

Toimittaja: Aviva Systems Biology
AVARP03045T100
AVIVAVARP03045T100EA 327 EUR
AVIVAVARP03045T100
Anti-CCNA2 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: CCNA2 (cyclin A2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR