Anti-CCDC173 Rabbit Polyclonal Antibody

Toimittaja: Aviva Systems Biology
ARP69256_P050
AVIVARP69256_P050EA 376 EUR
AVIVARP69256_P050
Anti-CCDC173 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: CCDC173 (coiled-coil domain containing 173)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR