Anti-TMEM121 Rabbit Polyclonal Antibody

Toimittaja: Aviva Systems Biology
ARP68966_P050
AVIVARP68966_P050EA 376 EUR
AVIVARP68966_P050
Anti-TMEM121 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: TMEM121 (transmembrane protein 121)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR