Anti-AFAP1L1 Rabbit Polyclonal Antibody

Toimittaja: Aviva Systems Biology
ARP67940_P050
AVIVARP67940_P050EA 376 EUR
AVIVARP67940_P050
Anti-AFAP1L1 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: AFAP1L1 (actin filament associated protein 1-like 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR