Anti-MYLPF Rabbit Polyclonal Antibody

Toimittaja: Aviva Systems Biology
ARP66217_P050
AVIVARP66217_P050EA 376 EUR
AVIVARP66217_P050
Anti-MYLPF Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: MYLPF (myosin light chain, phosphorylatable, fast skeletal muscle)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR