Anti-EIF2B2 Rabbit Polyclonal Antibody

Toimittaja: Aviva Systems Biology
ARP65664_P050
AVIVARP65664_P050EA 376 EUR
AVIVARP65664_P050
Anti-EIF2B2 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: EIF2B2 (eukaryotic translation initiation factor 2B, subunit 2 beta, 39kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR