Anti-EMR1 Rabbit Polyclonal Antibody

Toimittaja: Aviva Systems Biology
ARP64411_P050
AVIVARP64411_P050EA 376 EUR
AVIVARP64411_P050
Anti-EMR1 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: EMR1 (egf-like module containing, mucin-like, hormone receptor-like 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR