Anti-GPI Rabbit Polyclonal Antibody

Toimittaja: Aviva Systems Biology
ARP61088_P050
AVIVARP61088_P050EA 376 EUR
AVIVARP61088_P050
Anti-GPI Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: GPI (glucose-6-phosphate isomerase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR