Anti-PSMA3 Rabbit Polyclonal Antibody

Toimittaja: Aviva Systems Biology
ARP57743_P050
AVIVARP57743_P050EA 376 EUR
AVIVARP57743_P050
Anti-PSMA3 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: PSMA3 (proteasome (prosome, macropain) subunit, alpha type, 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR