Anti-MAPT Rabbit Polyclonal Antibody

Toimittaja: Aviva Systems Biology
ARP48103_P050
AVIVARP48103_P050EA 376 EUR
AVIVARP48103_P050
Anti-MAPT Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: MAPT (microtubule-associated protein tau)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR