Anti-KLRF1 Rabbit Polyclonal Antibody
Toimittaja: Aviva Systems Biology
ARP47263_P050
AVIVARP47263_P050EA 376 EUR
AVIVARP47263_P050
Anti-KLRF1 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: KLRF1 (killer cell lectin-like receptor subfamily F, member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human