Anti-IMPG2 Rabbit Polyclonal Antibody
Toimittaja: Aviva Systems Biology
ARP47238_P050
AVIVARP47238_P050EA 376 EUR
AVIVARP47238_P050
Anti-IMPG2 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: IMPG2 (interphotoreceptor matrix proteoglycan 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Order Now