Anti-UBE2L3 Rabbit Polyclonal Antibody

Toimittaja: Aviva Systems Biology
ARP43025_T100
AVIVARP43025_T100EA 327 EUR
AVIVARP43025_T100
Anti-UBE2L3 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: UBE2L3 (ubiquitin-conjugating enzyme E2L 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR