Anti-APOB Rabbit Polyclonal Antibody

Toimittaja: Aviva Systems Biology
ARP41726_P050
AVIVARP41726_P050EA 376 EUR
AVIVARP41726_P050
Anti-APOB Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: APOB (apolipoprotein B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR