Anti-A1BG Rabbit Polyclonal Antibody

Toimittaja: Aviva Systems Biology
ARP33810_P050 ARP33810_T100
AVIVARP33810_P050EA 376 EUR
AVIVARP33810_P050 AVIVARP33810_T100
Anti-A1BG Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: A1BG (alpha-1-B glycoprotein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR