Anti-PAX6 Rabbit Polyclonal Antibody

Toimittaja: Aviva Systems Biology
ARP32741_P050
AVIVARP32741_P050EA 376 EUR
AVIVARP32741_P050
Anti-PAX6 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: PAX6 (paired box 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR