Natriumkloridi BP, Ph. Eur., JP, USP, pure

Toimittaja: PanReac AppliChem

Synonyms: Saline, NaCl, Sea salt, Halite

141659.0416 141659.1211 141659.1214
APPC141659.0416EA 209 EUR
APPC141659.0416 APPC141659.1211 APPC141659.1214
Natriumkloridi BP, Ph. Eur., JP, USP, pure
Natriumkloridi
Kaava: NaCl
MDL numero: MFCD00003477
CAS-numero: 7647-14-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR