Murashige and skoog modified basal medium (with 2iP And IAA)
Toimittaja: Apollo Scientific
PMM702-10X1L PMM702-10L PMM702-50L
APOSPMM702-10X1LEA 0 EUR
APOSPMM702-10X1L APOSPMM702-10L APOSPMM702-50L
Murashige and skoog modified basal medium (with 2iP And IAA)
Soluviljelyyn liuokset
Contains macro/micronutrients as described by Murashige and Skoog, 1962. Modified with addition of 2iP, Adenine hemisulphate and IAA. Plant tissue culture testedFull specification given in appendix.
  • MFCD00240976