Murashige and skoog Medium with vitamins and sucrose
Toimittaja: Apollo Scientific
PMM5501B-10L PMM5501C-50L PMM5501D-100L
APOSPMM5501B-10LEA 51.4 EUR
APOSPMM5501B-10L APOSPMM5501C-50L APOSPMM5501D-100L
Murashige and skoog Medium with vitamins and sucrose
Soluviljelyyn liuokset
Modified version of product number PMM519, contains 30 g/L D-Sucrose. Plant Tissue Culture Tested. Soluble in water.