Diphenyliodonium hexafluorophosphate 98%

Toimittaja: Apollo Scientific
Vaara

PC8214-50G PC8214-10G PC8214-250G
APOSPC8214-50GEA 359 EUR
APOSPC8214-50G APOSPC8214-10G APOSPC8214-250G
Diphenyliodonium hexafluorophosphate 98%
Diphenyliodonium hexafluorophosphate
Kaava: C₁₂H₁₀F₆IP
Sulamispiste: 142 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00061398
CAS-numero: 58109-40-3
UN: 1759
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR