α,α,α-Trifluori-4-tolualdehydi 98%

Toimittaja: Apollo Scientific

PC7502-100G PC7502-25G
APOSPC7502-100GEA 64.3 EUR
APOSPC7502-100G APOSPC7502-25G
α,α,α-Trifluori-4-tolualdehydi 98%
α,α,α-Trifluori-4-tolualdehydi
Kaava: C₈H₅F₃O
MDL numero: MFCD00006952
CAS-numero: 455-19-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR