Tetrafluorosuccinyl chloride 95%

Toimittaja: Apollo Scientific
Vaara

PC6894-1G PC6894-5G
APOSPC6894-1GEA 168 EUR
APOSPC6894-1G APOSPC6894-5G
Tetrafluorosuccinyl chloride 95%
Tetrafluorosuccinyl chloride
Kaava: C₄Cl₂F₄O₂
Kiehumispiste: 85…87 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00013653
CAS-numero: 356-15-0
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR