[(S,S)-Ethylenebis(4,5,6,7-tetrahydroinden-1-yl)]difluorotitanium(IV)

Toimittaja: Apollo Scientific
Vaara

PC6741-100MG PC6741-25MG
APOSPC6741-100MGEA 199 EUR
APOSPC6741-100MG APOSPC6741-25MG
[(S,S)-Ethylenebis(4,5,6,7-tetrahydroinden-1-yl)]difluorotitanium(IV)
[(S,S)-Ethylenebis(4,5,6,7-tetrahydroinden-1-yl)]difluorotitanium(IV)

Precatalyst for the highly enantioselective imine hydrosilylation

Kaava: C₂₀H₂₄F₂Ti
Storage Temperature: Huoneenlämpö
MDL numero: MFCD01863622
CAS-numero: 178177-04-3
UN: 1759
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR