Perfluoroazelaic acid, Technical Grade

Toimittaja: Apollo Scientific
Vaara

PC6656-5G PC6656-1G
APOSPC6656-5GEA 103 EUR
APOSPC6656-5G APOSPC6656-1G
Perfluoroazelaic acid, Technical Grade
Perfluoroazelaic acid
Kaava: HO₂C(CF₂)₇CO₂H
Kiehumispiste: 215 °C/60mmHg
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00153274
CAS-numero: 23453-64-7
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR