Lithium difluoroacetate

Toimittaja: Apollo Scientific
Varoitus

PC5514-1G PC5514-5G
APOSPC5514-1GEA 103 EUR
APOSPC5514-1G APOSPC5514-5G
Lithium difluoroacetate
Lithium difluoroacetate
Kaava: C₂HF₂O₂·Li
Storage Temperature: Huoneenlämpö
MDL numero: MFCD03094294
CAS-numero: 74956-94-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR