Methyl 3-(1,1-difluoroethyl)benzoate 95%

Toimittaja: Apollo Scientific
Varoitus

PC53632-250MG PC53632-1G PC53632-5G
APOSPC53632-250MGEA 176 EUR
APOSPC53632-250MG APOSPC53632-1G APOSPC53632-5G
Methyl 3-(1,1-difluoroethyl)benzoate 95%
Methyl 3-(1,1-difluoroethyl)benzoate
Kaava: C₁₀H₁₀F₂O₂
MDL numero: MFCD30717684
CAS-numero: 878745-29-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR